ContactUsLinkComponent
Stericlin® sterilisation bags, gusseted, paper 60g/m², with STEAM indicator, 12.5 x 5 x 25.7 cm, DIN EN ISO 11607 1 * 1.000 items
Catalog # 129-1315
Stericlin® sterilisation bags, gusseted, paper 60g/m², with STEAM indicator, 12.5 x 5 x 25.7 cm, DIN EN ISO 11607 1 * 1.000 items
Catalog # 129-1315
Frequently Bought Together