ContactUsLinkComponent
Stericlin® sterilisation reel, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, width 20cm x le ngth 100m, DIN EN ISO 11607 1 * 2 Roll
Catalog # 129-1294
Stericlin® sterilisation reel, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, width 20cm x le ngth 100m, DIN EN ISO 11607 1 * 2 Roll
Catalog # 129-1294
Frequently Bought Together