ContactUsLinkComponent
Stericlin® sterilisation pouches, flat, paper 70g/m² with see-through film, with indicators for STEAM, EO and FORM, 30 x 57 cm, DIN EN ISO 11607 1 * 500 items
Catalog # 129-1257
Stericlin® sterilisation pouches, flat, paper 70g/m² with see-through film, with indicators for STEAM, EO and FORM, 30 x 57 cm, DIN EN ISO 11607 1 * 500 items
Catalog # 129-1257
Frequently Bought Together