ContactUsLinkComponent
Additional indicator strips (50) for Stericlin® BMS 1200 Batch Monitoring System, also includes 1 capsule, DIN EN 867-5 1 * 50 items
Catalog # 489-0128
Additional indicator strips (50) for Stericlin® BMS 1200 Batch Monitoring System, also includes 1 capsule, DIN EN 867-5 1 * 50 items
Catalog # 489-0128
Frequently Bought Together