ContactUsLinkComponent
Stericlin® thermal transfer film APX® FH+, for paper or plastic substrates, width 80mm x length 300m x core diameter 25.4mm 1 * 5 Roll
Catalog # VEPA3FSGZ815462
Stericlin® thermal transfer film APX® FH+, for paper or plastic substrates, width 80mm x length 300m x core diameter 25.4mm 1 * 5 Roll
Catalog # VEPA3FSGZ815462
Frequently Bought Together