ContactUsLinkComponent
Stericlin® container seal, loop into container lock and fold to seal, with a STEAM indicator on one half, and documentation fields o n the other half, 125x45mm (62.5x45mm folded), strip 2.5x70mm 1 * 500 items
Catalog # VEPA3FCZB460110
Stericlin® container seal, loop into container lock and fold to seal, with a STEAM indicator on one half, and documentation fields o n the other half, 125x45mm (62.5x45mm folded), strip 2.5x70mm 1 * 500 items
Catalog # VEPA3FCZB460110
Frequently Bought Together