ContactUsLinkComponent
Stericlin® tray liner, made of white crepe paper, to protect wrapping materials from mesh trays, dimensions 58x35cm, pre-formed for tray size 48x25cm 1 * 500 items
Catalog # VEPA3FTSZ420104
Stericlin® tray liner, made of white crepe paper, to protect wrapping materials from mesh trays, dimensions 58x35cm, pre-formed for tray size 48x25cm 1 * 500 items
Catalog # VEPA3FTSZ420104
Frequently Bought Together