CHLOROFORM CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Catalog # 6754-25
Supplier: MACRON AVANTOR BRAND
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water jet pump
Supplier: Brand
VWR®, Disposable Micro Weighing Dishes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
HPLC columns, LiChrospher®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals