ContactUsLinkComponent
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 3/4 Sanitary Flange x 3/8” Hosebarb 1 * 1 items
Catalog # PENDPREPS-N-5-038
Supplier:  PENDOTECH
PendoTech Single-Use Pressure Sensor, Non-Sterile, Polysulfone, 3/4 Sanitary Flange x 3/8” Hosebarb 1 * 1 items
Catalog # PENDPREPS-N-5-038
Supplier:  PENDOTECH