Orders LinkContactUsLinkComponent
Nalgene™ Staining Boxes
Nalgene™ Staining Boxes
Replacement plug for 5705-1010 and 2020, PMP, —, — mm
Catalog # NALG715705-0001
   Low Manufacturing Impact
Nalgene™ Staining Boxes
Catalog # NALG715705-0001
   Low Manufacturing Impact
Nalgene™ Staining Boxes
Nalgene™ Staining Boxes
Replacement plug for 5705-1010 and 2020, PMP, —, — mm
Price
SFr. 32.40
Log in for availability
1 * 1 ST
Product Details & Documents
Stain, fix and handle fragile electrophoresis gels and membranes with less chance of damage than conventional methods.
  • Polymethylpentene box and polypropylene copolymer cover
  • Useful over a –70 to +70 °C (–94 to +158 °F) temperature range
  • Good chemical compatibility with acids, bases and select organic solvents for a versatile range of protocols
  • LDPE plug in bottom for easy drainage of liquid with minimal handling of gel and provides a leakproof seal
  • Plug won't interfere with cover or contents of boxes on a benchtop or shaker
  • Stackable, with a handle on each end for safe, easy carrying
  • Microwavable and easy to clean
Thermo Scientific™ Nalgene™ staining box is equipped with a bottom drain allowing you to change solution without disturbing the box's other fragile contents. Box holds 189×189 mm (4,8×4,8") gel along with 500 ml of liquid.
Specifications
  • Capacity:
  • Description:
    Replacement plug for 5705-1010 and 2020
  • Material:
    PMP
  • Seq:
    02
  • W×D×H:
    — mm
  • Cat. No.:
    NALG715705-0001
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact