ContactUsLinkComponent
Grinding Cylinder 6,35 x 12,7 mm Stainless Steel Lysing Matrix Tubes are ideal for grinding, lysis and homogenization of your most resistant samples The stainless steel threaded cap provides a leak-proof closure without the energy-robbing alternativ 1 * 3 items
Catalog # 116992003.
Supplier:  MP Biomedicals
Grinding Cylinder 6,35 x 12,7 mm Stainless Steel Lysing Matrix Tubes are ideal for grinding, lysis and homogenization of your most resistant samples The stainless steel threaded cap provides a leak-proof closure without the energy-robbing alternativ 1 * 3 items
Catalog # 116992003.
Supplier:  MP Biomedicals
Frequently Bought Together