Orders LinkContactUsLinkComponent
Tablet counting triangles, SteriWare®
Tablet counting triangles, SteriWare®
Non-sterile, White, 230×230×13 mm
Catalog # SAMP8066A-1
Supplier:   Sampling Systems
Tablet counting triangles, SteriWare®
Catalog # SAMP8066A-1
Supplier:  Sampling Systems
Tablet counting triangles, SteriWare®
Tablet counting triangles, SteriWare®
Non-sterile, White, 230×230×13 mm
Price
SFr. 194.00
Log in for availability
1 * 30 ST
Product Details & Documents
The SteriWare® counting triangles are ideal for counting round tablets. They are manufactured and packed in a cleanroom and so can be used straight from the bag.
  • Made from FDA conforming PS
  • BSE/TSE free
  • Suitable for left handed and right handed operators
  • With an embossed scale to aid operation
Zertifizierung: Conforms to FDA, EU 10/2011 and EC 1935/2004. Sterility Assurance Level (SAL) of 10⁻⁶.
Verpackung: Individually bagged in a heat-sealed laminate bag.
Specifications
  • Colour:
    White
  • Description:
    Non-sterile
  • Seq:
    02
  • W×D×H:
    230×230×13 mm