Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
WASTE BAG BD 110L (10 ROLLS OF 20) 1 * 200 items
Catalog # GMEQOJD003
Supplier:
GM EQUIPEMENT
Expand image
WASTE BAG BD 110L (10 ROLLS OF 20) 1 * 200 items
Catalog # GMEQOJD003
Supplier:
GM EQUIPEMENT
Frequently Bought Together
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
Gloves, nitrile/neoprene, Microflex® 93-260
Supplier: Ansell
VWR®, Nitrile Gloves
Supplier: VWR Collection
Lead-free steam indicator tapes, Comply™
Supplier: 3M MEDICAL
VWR®, Laboratory Markers
Supplier: VWR Collection
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR® Nitrile Light Long, Nitrile Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection