ContactUsLinkComponent
Cup horn offers indirect sonication to keep samples sterile and/or to process samples under 200ul. Only for use with Q125 Sonicator. 1 * 1 items
Catalog # QSON4608
Supplier:  QSONICA LLC
Cup horn offers indirect sonication to keep samples sterile and/or to process samples under 200ul. Only for use with Q125 Sonicator. 1 * 1 items
Catalog # QSON4608
Supplier:  QSONICA LLC
Frequently Bought Together