ContactUsLinkComponent
Turbidity Standard, Calibration value of 1010 FNU for ProDSS and EXO sensors, calibration value of 1000 for YSI 6136 sensor 1 * 1 items
Catalog # YSIA607400
Supplier:  YSI ENVIRONMENTAL
Turbidity Standard, Calibration value of 1010 FNU for ProDSS and EXO sensors, calibration value of 1000 for YSI 6136 sensor 1 * 1 items
Catalog # YSIA607400
Supplier:  YSI ENVIRONMENTAL
Frequently Bought Together