ZOBELL SOLUTION, ORP CALIBRATOR 1 * 250 mL
Catalog # YSIA061321
Supplier: YSI ENVIRONMENTAL
Frequently Bought Together
VWR®, Redox (ORP) Standards
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Murashige and skoog basal mixture
Supplier: Apollo Scientific