Order Entry
Switzerland
Orders LinkContactUsLinkComponent
4-Hydroxycinnamic acid 98%

4-Hydroxycinnamic acid 98%

Bottle, 100 g
Catalog # APOSOR21362-100G
Supplier:   Apollo Scientific
4-Hydroxycinnamic acid 98%
Catalog # APOSOR21362-100G
Supplier:  Apollo Scientific
CAS Number:  7400-08-0
4-Hydroxycinnamic acid 98%

Bottle, 100 g

CAS Number:  7400-08-0
Price
SFr. 65.90
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  p-Coumaric acid, p-Cumaric acid, 3-(4-Hydroxyphenyl)acrylic acid, p-Hydroxy-Cinnamic Acid
  • CAS Number:7400-08-0
  • Formula:C₉H₈O₃
  • MDL Number:MFCD00004399
  • Molecular Weight:164.16 g/mol
  • Boiling Pt:346 °C
  • Melting Pt:212…214 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications