ContactUsLinkComponent
YMC-TRIART C8 PREPARATIVE COLUMNS, S-5, 120Å, 10 X 150 MM 1 * 1 items
Catalog # YMCATO12S05-1510WT
Supplier:  YMC
YMC-TRIART C8 PREPARATIVE COLUMNS, S-5, 120Å, 10 X 150 MM 1 * 1 items
Catalog # YMCATO12S05-1510WT
Supplier:  YMC