Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Mailing boxes, EPS

Protective cardboard outer box, for 3 L box, 244×259×162 mm, 242×240×133 mm
  DAKL520006
 :   DAKLAPACK
Mailing boxes, EPS
  DAKL520006
 :  DAKLAPACK
Mailing boxes, EPS

Protective cardboard outer box, for 3 L box, 244×259×162 mm, 242×240×133 mm

 
SFr. 24.70
   
1 * 1 ST
 

Made up of expanded polystyrene (EPS).

  • Non transparent
  • Can be used for cooling elements, gel packs and perfect for dry ice

Suitable for refrigerated transport of biological substances, pharmaceuticals, and blood products.

 
  • Description:
    Protective cardboard outer box, for 3 L box
  • Seq:
    04
  • W×D×H ext.:
    244×259×162 mm
  • W×D×H int.:
    242×240×133 mm