Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Human Recombinant FGF-basic 147 (from <i>E. coli</i>)

Human Recombinant FGF-basic 147 (from E. coli)

Vial, 50 µG
Catalog # SHBT100-28-50UG
Human Recombinant FGF-basic 147 (from <i>E. coli</i>)
Catalog # SHBT100-28-50UG
CAS Number:  
Human Recombinant FGF-basic 147 (from <i>E. coli</i>)
Human Recombinant FGF-basic 147 (from E. coli)

Vial, 50 µG

CAS Number:  
Price
SFr. 182.00
Log in for availability
1 * 50 µG
Product Details & Documents

Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Degredation of the full length FGF-basic N-terminus results in a truncated FGF-basic 147aa protein, when the protein is isolated from biological sources. The N-terminus extensions influence the localization of FGF-basic within the cell, but do not affect the biological activity of FGF-basic. Therefore, there are no detectable differences in biological activity between the full length FGF-basic 154aa and the truncated FGF-basic 147aa recombinant proteins.

  • High Quality
  • Low Endotoxin
  • Affordable

A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  

Caution: For research use only.

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    50 µG
  • Protease-free:
    N
  • Purity:
    ≥95%
  • Source:
    E. coli
  • Molecular weight:
    16,5 kDa
  • Nuclease-free:
    N
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Protein function:
    Cytokine
  • Gene ID:
    P09038
  • Formulation:
    10 mM sodium phosphate, 75 mM sodium chloride, pH 7,5
  • Endotoxin-free:
    N
  • Tested applications:
    Bioassay
  • Species:
    Human
  • Protein/peptide name:
    FGF-basic 147
  • Protein synonyms:
    Prostatropin|HBGF-2|FGF2
  • Endotoxin level:
    Low
  • Grade:
    RUO
  • Sequence:
    MPALPEDGGS GAFPPGHFKD PKRLYCKNGG FFLRIHPDGR VDGVREKSDP HIKLQLQAEE RGVVSIKGVC ANRYLAMKED GRLLASKCVT DECFFFERLE SNNYNTYRSR KYTSWYVALK RTGQYKLGSK TGPGQKAILF LPMSAKS
  • Biological activity:
    Bioactivity assay: NR6R-3T3 cell proliferation, ED50 ≤5 ng/ml, Bioactivity ≥2,0×10⁵ units/mg
  • Shipping temperature:
    RT
  • Cat. No.:
    SHBT100-28-50UG
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Storage conditions:
    –20 °C
  • Carrier-free:
    Y
  • Conjugation:
    Unconjugated
  • Animal-free:
    N

Specifications