Rat Recombinant FGF-basic (from E. coli)
Vial, 10 µG
Catalog # SHBT300-02-10UG
Supplier: Shenandoah Biotechnology
Rat Recombinant FGF-basic (from <i>E. coli</i>)
Catalog # SHBT300-02-10UG
Supplier: Shenandoah Biotechnology
CAS Number:
Jump to:
Product Details & Documents
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic binds heparin in order to signal through fibroblast growth factor (FGF) receptor tyrosine kinases.
- High Quality
- Low Endotoxin
- Affordable
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
Achtung! Bitte beachten: For research use only.
Specifications
Specifications
- Pack type:Vial
- Pk:10 µG
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:16,4 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Protein function:Cytokine
- Gene ID:P13109
- Formulation:10 mM sodium phosphate, 50 mM sodium chloride, pH 7,5
- Endotoxin-free:N
- Tested applications:Bioassay
- Species:Rat
- Protein/peptide name:FGF-basic
- Protein synonyms:Prostatropin|HBGF-2|FGF2
- Endotoxin level:Low
- Grade:RUO
- Sequence:MPALPEDGGG AFPPGHFKDP KRLYCKNGGF FLRIHPDGRV DGVREKSDPH VKLQLQAEER GVVSIKGVCA NRYLAMKEDG RLLASKCVTE ECFFFERLES NNYNTYRSRK YSSWYVALKR TGQYKLGSKT GPGQKAILFL PMSAKS
- Biological activity:Bioactivity assay: NR6R 3T3 proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1,0×10⁶ units/mg
- Shipping temperature:RT
- Cat. No.:SHBT300-02-10UG
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Storage conditions:–20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:N



