Human Recombinant FGF-Basic 154 (from E. coli)
Vial, 100 µG
Catalog # SHBT100-146-100UG
Supplier: Shenandoah Biotechnology
Human Recombinant FGF-Basic 154 (from <i>E. coli</i>)
Catalog # SHBT100-146-100UG
Supplier: Shenandoah Biotechnology
CAS Number:
Jump to:
Product Details & Documents
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis. FGF-basic also has cardioprotective functions during heart injury. FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state. Recombinant FGF-basic 154 is the full length FGF-basic protein encoded by the human FGF-2 gene. FGF-basic 154 is the most popular tissue culture product at Shenandoah Biotechnology, Inc. There are no detectable differences in biological activity between FGF-basic 154 and the truncated FGF-basic 147 proteins.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Documents
Literature reference - PubMed ID 26225855
Additional Product Details
Specifications
Specifications
- Pack type:Vial
- Pk:100 µG
- Protease-free:N
- Purity:≥95%
- Source:E. coli
- Molecular weight:17,3 kDa
- Nuclease-free:N
- Protein/peptide type:Recombinant
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Protein function:Cytokine
- Gene ID:P09038
- Formulation:10 mM Sodium phosphate, 75 mM Sodium chloride, pH 7,5
- Endotoxin-free:N
- Tested applications:Bioassay
- Species:Human
- Protein/peptide name:FGF-Basic 154
- Protein synonyms:Prostatropin|HBGF-2|FGF2
- Endotoxin level:Low
- Grade:RUO
- Sequence:MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY TSWYVALKRT GQYKLGSKTG PGQKAILFLP MSAKS
- Biological activity:Bioactivity Assay: NR6R-3T3 Proliferation, ED₅₀ ≤0,5 ng/ml, Bioactivity ≥2,0×10⁶ units/mg
- Shipping temperature:RT
- Cat. No.:SHBT100-146-100UG
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Storage conditions:–20 °C
- Carrier-free:Y
- Conjugation:Unconjugated
- Animal-free:N



