ContactUsLinkComponent
AL 251 - orange Flash Light and buzzer combination - You can connect the LED / buzzer combination to the potential-free contact of the alarm boxfor visual alarm in case of limit violations. This device can be used as an accessory to the EBI 25 system 1 * 1 items
Catalog # EBRO1340-6233
Supplier:  EBRO
undefined
AL 251 - orange Flash Light and buzzer combination - You can connect the LED / buzzer combination to the potential-free contact of the alarm boxfor visual alarm in case of limit violations. This device can be used as an accessory to the EBI 25 system 1 * 1 items
Catalog # EBRO1340-6233
Supplier:  EBRO
Frequently Bought Together