ContactUsLinkComponent
RNASCOPE VS UNIVERSAL AP REAGENT KIT IS A RNA IN SITU HYBRIDIZATION (ISH) SINGLE-PLEX,CHROMOGENIC -RED ASSAY DESIGNED FOR USE WITH THE DISCOVERY ULTRA AUTOMATED IHC/ISH SLIDESTAINING SYSTEMS BY VENTANA MEDICAL SYSTEMS. 1 * 1 KIT
  ADVD323250
RNASCOPE VS UNIVERSAL AP REAGENT KIT IS A RNA IN SITU HYBRIDIZATION (ISH) SINGLE-PLEX,CHROMOGENIC -RED ASSAY DESIGNED FOR USE WITH THE DISCOVERY ULTRA AUTOMATED IHC/ISH SLIDESTAINING SYSTEMS BY VENTANA MEDICAL SYSTEMS. 1 * 1 KIT
  ADVD323250
Frequently Bought Together