ContactUsLinkComponent
Multi-purpose container. Nominal volume 15 Liters. Complete with a swing carrying handle. Height 285 mm. 267 mm. 304 stainless steel. Toogle clamps and O-Ring seals to be ordered separately. 1 * 1 items
Catalog # SAMPA434-15
Supplier:  Sampling Systems
Multi-purpose container. Nominal volume 15 Liters. Complete with a swing carrying handle. Height 285 mm. 267 mm. 304 stainless steel. Toogle clamps and O-Ring seals to be ordered separately. 1 * 1 items
Catalog # SAMPA434-15
Supplier:  Sampling Systems
Frequently Bought Together