ContactUsLinkComponent
Wash bottles, Capacity 1 Liter. Narrow-mouth bottles, made from PE-LD coloured red. Screw cap and spray tube from PP natural. Thread GL 32. Ø mm 92. Height mm 221. 1 * 5 items
Catalog # VITL132803
Supplier:  VITLAB
Wash bottles, Capacity 1 Liter. Narrow-mouth bottles, made from PE-LD coloured red. Screw cap and spray tube from PP natural. Thread GL 32. Ø mm 92. Height mm 221. 1 * 5 items
Catalog # VITL132803
Supplier:  VITLAB
Frequently Bought Together