Rabbit polyclonal antibody raised against partial recombiant human EML4. Application: WB-Ce,IHC-P,IF 1 * 100 µl
ABNOPAB31053
: Abnova
Rabbit polyclonal antibody raised against partial recombiant human EML4. Application: WB-Ce,IHC-P,IF 1 * 100 µl
ABNOPAB31053
: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection