Mouse monoclonal antibody raised against partial recombinant human CDH1. Application: WB-Ce,IHC-P 1 * 100 µl
ABNOMAB15671
: Abnova
Mouse monoclonal antibody raised against partial recombinant human CDH1. Application: WB-Ce,IHC-P 1 * 100 µl
ABNOMAB15671
: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection