Tri-Potassium citrate monohydrate EMPROVE ESSENTIAL Ph Eur,BP,USP,FCC,E 332 1 * 1 kg
Catalog # 1.04956.1000
Supplier: MERCK PRODUCTION CHEMICALS
Tri-Potassium citrate monohydrate EMPROVE ESSENTIAL Ph Eur,BP,USP,FCC,E 332 1 * 1 kg
Catalog # 1.04956.1000
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Sodium dihydrogen phosphate dihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Magnesium oxide light 98.0-100.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Tin(II) chloride dihydrate ≥98%, AnalaR NORMAPUR® analytical reagent (max. 0.05 ppm Hg)
Supplier: VWR Chemicals
Potassium dihydrogen phosphate, crystallised, EMPROVE® ESSENTIAL Ph. Eur., BP, JPC, NF, E340, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Sodium chloride ≥98%, TECHNICAL
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium chloride ≥97%, TECHNICAL
Supplier: VWR Chemicals
Potassium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals