Bag Precision Clean II, made of clear amine-free Low Density PolyEthylene, suitable for use in cleanrooms, manufactured and packed in an ISO 4 cleanroom, thickness 4mil (100 micron) per layer, width/opening 6 (152mm), length 9 (229mm), 2000 pieces 1 * 2.000 items
Catalog # FISH11028
Supplier: FISHER CONTAINER
Frequently Bought Together