Tanks, rectangular, LDPE, LaboPlast®
Lid, LDPE, for rectangular tanks, 50 L
Catalog # BURK3440-0501
Supplier: BURKLE
Jump to:
Product Details & Documents
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
- Linear LDPE, transparent
- Rupture-proof
Specifications
- Description:Lid, LDPE, for rectangular tanks, 50 L
- Seq:23
Accessories
Product Information
- Catalog #AvailabilityPrice
- Container thread ¾" for cylindrical containersContainer thread ¾" for cylindrical containersBURK3440-0034Log in for availability1 * 1 ST - SFr. 202.00