Order Entry
Switzerland
Orders LinkContactUsLinkComponent
High content cell imaging system, micro confocal, ImageXpress

High content cell imaging system, micro confocal, ImageXpress

Micro confocal environment control and transmitted light with phase contrast combination including 20× Ph1 S plan fluor ELWD adm 0,45 N.A objective
Catalog # MLDV5041309
Supplier:   Molecular Devices
High content cell imaging system, micro confocal, ImageXpress
Catalog # MLDV5041309
Supplier:  Molecular Devices
High content cell imaging system, micro confocal, ImageXpress
High content cell imaging system, micro confocal, ImageXpress

Micro confocal environment control and transmitted light with phase contrast combination including 20× Ph1 S plan fluor ELWD adm 0,45 N.A objective

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

The ImageXpress® Micro-confocal high content imaging system helps to expand the boundaries of research with a combination of speed, sensitivity and flexibility, giving you higher quality images, at a faster throughput, with more powerful analysis.

  • Quickly and easily switch between widefield and several confocal disc configurations, including an exclusive slit option, to capture images at high throughput speeds
  • Image more than 160000 wells per day in confocal mode
  • Excellent image quality via the unique optical path technology
  • Large field of view enables whole-well imaging
  • Tackle a wide range of assays with water, oil and air objectives ranging from 1× to 100×
  • Expand your research capabilities with water immersion objectives, transmitted light, phase contrast optics, on-board liquid handling, and environmental control options

The ImageXpress® Micro-confocal system lets you capture research quality images of a vast range of assays, including whole organisms, thick tissues, 3-D spheroid assays, and cellular or intracellular events - at a speed you would only expect from widefield screening. Benefit from a wide range of imaging modes, including phase contrast and brightfield label-free imaging, fluorescence, widefield, colorimetric, and confocal imaging, to generate publication quality images at high throughput. The system is designed to be exceptionally versatile with user exchangeable filter cubes, a selection of confocal geometries to match your exact assay requirements, on board fluidics and environmental control, the option to benefit from laser illumination, and much more.

Combined with MetaXpress® high content image acquisition and analysis software, the ImageXpress® Micro-confocal system provides you with a complete, customisable, high throughput screening solution to discover your landmark scientific breakthrough.

Delivery information: Supplied with a 10× plan-apochromatic 0,45 N.A objective, MetaXpress® imaging and analysis software, Windows® 7 64-bit acquisition computer with license to run MetaXpress® powercore and MDCstore database. Confocal screening option includes eight position filter wheel, five position dichroic wheel and dual disk unit.

Specifications
  • Description:
    Micro confocal environment control and transmitted light with phase contrast combination including 20× Ph1 S plan fluor ELWD adm 0,45 N.A objective
  • Seq:
    05
Complementary Products

Product Information

  • Catalog #
    Description
    Pk
    Serum format
    Sterility
    Packed
    Sterile
    Working volume
    Well volume
    Colour
    Recommended working volume
    Availability
    Price
  • 30009-2
    Caspase-3 DEVD-R110 Fluorometric HTS Assay Kit, 10 ml
    1 KIT
    Log in for availability
    1 * 1 KIT - SFr. 354.00

  • 30026.
    Calcein-AM Cell Viability Assay kit, 1000 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - SFr. 501.00

  • 100.103.010
    GrowDex®, 10 ml syringe
    1
    Log in for availability
    1 * 1 ST - SFr. 348.00

  • S1810-500
    Foetal bovine serum, South American origin
    Frozen
    15
    Log in for availability
    1 * 500 mL - SFr. 330.00

  • 30002.
    Viability/Cytotoxicity Assay kit for Animal Live & Dead Cells, 300 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - SFr. 528.00

  • 30029.
    NucView™ 488 Caspase-3 Assay Kit for live cells, 100 assays
    1 KIT
    Log in for availability
    1 * 1 KIT - SFr. 758.00

  • 735-0523
    Microlite™ 1+ strip, 1×12 per strip
    Log in for availability
    1 * 320 ST - SFr. 252.00

  • CORN3904BC
    20/bag
    96-well microplate, clear flat bottom, tissue culture treated, with lid
    75 - 200 µl
    21
    360 µl
    Black
    Log in for availability
    1 * 100 ST - SFr. 1’380.00

  • 734-2327
    1/bag
    0,075 - 0,20 ml
    0,33 cm²
    +
    17
    Clear
    Log in for availability
    1 * 100 ST - SFr. 305.00

  • 734-2097
    200 µl
    96-MicroWell™ plate, flat bottomed wells, with lid
    Transparent
    Log in for availability
    1 * 50 ST - SFr. 271.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.