Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Human CD38 Protein (ECD, His Tag), HPLC-verified 1 * 50 µG
Catalog # SINO10818-H08H50UG
Supplier:
Sino Biological
Certificates
Expand image
Recombinant Human CD38 Protein (ECD, His Tag), HPLC-verified 1 * 50 µG
Catalog # SINO10818-H08H50UG
Supplier:
Sino Biological
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Test Tubes without Rim
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Phosphate Buffered Saline (PBS) with 0,05% Tween®20
Supplier: VWR Chemicals
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF