ContactUsLinkComponent
Precision thermometer made of Quartz-Glass, with ground joint 14,5/23, -10+250:0,5°C, upper part 350xc.9mm, built-in length 75mm, en closed scale, cap. colourless, gallium filled, pin-top finish, suitable for government verification 1 * 1 items
Catalog # AMARQ147032-GA
Supplier:  Amarell
Precision thermometer made of Quartz-Glass, with ground joint 14,5/23, -10+250:0,5°C, upper part 350xc.9mm, built-in length 75mm, en closed scale, cap. colourless, gallium filled, pin-top finish, suitable for government verification 1 * 1 items
Catalog # AMARQ147032-GA
Supplier:  Amarell
Frequently Bought Together