ContactUsLinkComponent
Amarell-Max.-therm. for swinging, quartz-glass, +395+750 :2°C, enclosed scale, cap. round, colourless, gallium filled, 300x8-9mm, pi n-top finish and button, manufactured for reading after cooling down to 23°C, suitable for government verification 1 * 1 items
Catalog # AMARL38146-GA
Supplier:  Amarell
Amarell-Max.-therm. for swinging, quartz-glass, +395+750 :2°C, enclosed scale, cap. round, colourless, gallium filled, 300x8-9mm, pi n-top finish and button, manufactured for reading after cooling down to 23°C, suitable for government verification 1 * 1 items
Catalog # AMARL38146-GA
Supplier:  Amarell
Frequently Bought Together