ContactUsLinkComponent
Soil therm., British type, similar specification, right angle, -7+38:0,5°C, solid stem, white backed, red special liquid, round bulb with diameter 11,5-14,0mm, built-in length 25mm (MK2A), lower part 50mm, suitable for government verification 1 * 1 items
Catalog # AMARK188600-FL
Supplier:  Amarell
Soil therm., British type, similar specification, right angle, -7+38:0,5°C, solid stem, white backed, red special liquid, round bulb with diameter 11,5-14,0mm, built-in length 25mm (MK2A), lower part 50mm, suitable for government verification 1 * 1 items
Catalog # AMARK188600-FL
Supplier:  Amarell
Frequently Bought Together