ContactUsLinkComponent
Thermometer similar to BS 593, F150C/76, -20+150:1C, solid stem, white backed, red special liquid, 322mm long, 76mm immersion, dura ble pigment, suitable for government verification 1 * 1 items
Catalog # AMARA403404-FL
Supplier:  Amarell
Thermometer similar to BS 593, F150C/76, -20+150:1C, solid stem, white backed, red special liquid, 322mm long, 76mm immersion, dura ble pigment, suitable for government verification 1 * 1 items
Catalog # AMARA403404-FL
Supplier:  Amarell
Frequently Bought Together