ContactUsLinkComponent
Thermometer similar to BS 593, F125C/100, 72+126:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 395mm long, 100mm immersion, durable pigment, null
Katalognr. AMARA403386-CB
Supplier:  Amarell
Thermometer similar to BS 593, F125C/100, 72+126:0,2°C, solid stem, white backed, capillary tube specially coated inside, with non- wetting blue special liquid, 395mm long, 100mm immersion, durable pigment, null
Katalognr. AMARA403386-CB
Supplier:  Amarell
Frequently Bought Together