ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 47 F, white backed, 137,5+142,5:0,1F, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300670-CB
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 47 F, white backed, 137,5+142,5:0,1F, capillary tube specially coated inside, with non-we tting blue special liquid, total immersion, max.310x6-8mm, durable pigment, with ice point scale, null
Catalog # AMARA300670-CB
Supplier:  Amarell
Frequently Bought Together