ContactUsLinkComponent
Sweet wort hydrometer, 0-10:0,1%mas, with therm. +10+30:0,2C with red or blue special liquid, with correction scale PTB 1626/30, re ading above, ref. temp. 20C, 370mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH847380-FL
Supplier:  Amarell
Sweet wort hydrometer, 0-10:0,1%mas, with therm. +10+30:0,2C with red or blue special liquid, with correction scale PTB 1626/30, re ading above, ref. temp. 20C, 370mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH847380-FL
Supplier:  Amarell
Frequently Bought Together