Beer boiling hydrometer, 15-20:0,1%mas, with therm. +5+25:0,2C with red or blue special liquid, correction scale PTB I 3685/29, rea ding above, ref. temp. 20C, 400mm long, suitable for government calibration 1 * 1 items
Katalognr. AMARH847316-FL
Supplier: Amarell
Frequently Bought Together