ContactUsLinkComponent
Beer boiling hydrometer, 10-15:0,1%mas, with therm. +5+25:0,2C with red or blue special liquid, correction scale PTB I 3685/29, rea ding above, ref. temp. 20C, 400mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH847314-FL
Supplier:  Amarell
Beer boiling hydrometer, 10-15:0,1%mas, with therm. +5+25:0,2C with red or blue special liquid, correction scale PTB I 3685/29, rea ding above, ref. temp. 20C, 400mm long, suitable for government calibration 1 * 1 items
Catalog # AMARH847314-FL
Supplier:  Amarell
Frequently Bought Together