Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Compact burettes, BLAUBRAND®

10 ml, 0,02 ml, 0,02 ± ml
Catalog # 612-7162
Supplier:   Brand
Compact burettes, BLAUBRAND®
Catalog # 612-7162
Supplier:  Brand
Compact burettes, BLAUBRAND®
Compact burettes, BLAUBRAND®

10 ml, 0,02 ml, 0,02 ± ml

Price
SFr. 127.00
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate 3.3 glass, amber, class AS.

  • Removable PTFE stopcock, fast to dismantle and easy to clean
  • Simple repair - all individual components replaceable
  • Calibrated to deliver (TD, Ex)

 : DIN EN ISO 385, DE-M marking

 : On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.

 : Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer.

Specifications
  • Capacity:
    10 ml
  • Seq:
    01
  • Division:
    0,02 ml
  • Tolerance:
    0,02 ± ml