ContactUsLinkComponent
BOTTLE REAGENT 2000ML -1/+1,5 BAR GL45 1 * 6 items
Catalog # KAVA1632414491950
Supplier:  Kavalier
BOTTLE REAGENT 2000ML -1/+1,5 BAR GL45 1 * 6 items
Catalog # KAVA1632414491950
Supplier:  Kavalier
Frequently Bought Together