Microarray molds kits, T-Sue™
Product Details & Documents
T-Sue™ Microarray mold, enabling tissues from many donor blocks to be arrayed on a single slide.
- Can be reused hundreds of times without losing its flexibility
- Withstands temperatures between –100 to +250 °C
- View many different samples on the same slide
- Array mold is designed to be simple and easy to use
The precast T-Sue™ Microarray paraffin blocks allow users to prepare tissue arrays without needing any specialized equipment. Construct a tissue array block in minutes, simply by punching the donor tissue cores and inserting them into the pre-made paraffin recipient block. No specific technical training or experience is needed.
They are ready to accept cores from a donor block, and offer a consistent core diameter. The cassette is permanently bonded to the block.T-Sue™ punch needles are used for filling recipient tissue microarray paraffin blocks. Internal stylet operates like a retractable pen. Effortless ejection of core specimens.
Tissues can be analyzed in the same conditions enhancing the efficiency of the research. The T-Sue™ Microarray molds allow users to perform TMAs faster while giving excellent results.
Lieferinformationen: Each kit includes ONE T-Sue™ array mold and FOUR punch needles with stylet allowing easy removal of tissue cores for insertion.
Specifications
- Description:T-Sue™ microarray mold, 15 mold cores, 4 mm core diameter
- Seq:01