Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Low temperature covers

Low temperature covers

Low temperature cover, plexiglass, 4 blocks, 216×216×41 mm
Catalog # OHAU30400180
Supplier:   OHAUS
Low temperature covers
Catalog # OHAU30400180
Supplier:  OHAUS
Low temperature covers
Low temperature covers

Low temperature cover, plexiglass, 4 blocks, 216×216×41 mm

Price
SFr. 121.00
Log in for availability
1 * 1 ST
Product Details & Documents

Plexiglass cover reduces air flow for additional temperature stability in low temperature applications.

  • Two sides are open 6,4 mm
Specifications
  • Description:
    Low temperature cover, plexiglass
  • Seq:
    02
  • W×D×H:
    216×216×41 mm
  • For:
    4 blocks
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.