Modular heating blocks
Single block, Ø 13,9 mm, 12/13 mm test tubes, 16, 48,4 mm, 95×76×51 mm
OHAU30400165
: OHAUS
Modular blocks are constructed from a solid anodized aluminium block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Caution: To avoid possible electrical hazard, do not fill well or block with water or other fluids. Units are designed as a dry bath/incubator.
- No. of Holes:16
- Description:Single block
- Seq:016
- W×D×H:95×76×51 mm
- Depth:48,4 mm
- Well size:Ø 13,9 mm
- For:12/13 mm test tubes



