Sandbaths for modular heating blocks
Sand bath for six-block heater, 287×147×64 mm
Catalog # OHAU30400176
Supplier: OHAUS
Jump to:
Product Details & Documents
Ideal for irregular shaped vessels.
- Stainless steel construction for superior corrosion resistance
- Designed to hold sand, stainless steel shot or non-volatile liquids
Specifications
- Description:Sand bath for six-block heater, 287×147×64 mm
- Seq:04
Accessories
Product Information
- Catalog #AvailabilityPrice
- Sand, 0,5 kg bag, for sand bath
Product Information
- Catalog #AvailabilityPrice
- Stainless steel shot, 0,5 kg, for sand bathStainless steel shot, 0,5 kg, for sand bathOHAU30400178Log in for availability1 * 1 ST - SFr. 103.50
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



