pH Buffer Solution pH 4.01 ± 0.02 ,25°C (Coloured) 1 * 500 mL
Catalog # REGETB401
Supplier: REAGECON
pH Buffer Solution pH 4.01 ± 0.02 ,25°C (Coloured) 1 * 500 mL
Catalog # REGETB401
Supplier: REAGECON
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Malachite green solution Reag. Ph. Eur. 1050501
Supplier: VWR Chemicals
DUKE STD 300NM 1 * 15 mL
Supplier: MARKET SOURCE PART PROCESS
Ferroin (1,10-phenanthrolineferrous sulphate complex) 0.025 M in aqueous solution
Supplier: Thermo Fisher Scientific
NATRIUMACETAT TRIHYDRAT 1 * 2 g
Supplier: Merck
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
Dichlobenil
Supplier: EHRENSTORFER
Saflufenacil-N,N-desmethyl 1 * 10 mg
Supplier: EHRENSTORFER
Colour Standard Gardner 5 1 * 100 mL
Supplier: REAGECON
pH buffer, single-use, SINGLET™
Supplier: Hach