Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Rat EGF

10 µG
Catalog # APOSBITP2077-10UG
Supplier:   Apollo Scientific
Rat EGF
Catalog # APOSBITP2077-10UG
Supplier:  Apollo Scientific
CAS Number:  
Rat EGF

10 µG

CAS Number:  
Price
SFr. 936.00
Log in for availability
1 * 10 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    10 µG
  • Cat. No.:
    APOSBITP2077-10UG
  • Protein/peptide name:
    EGF
  • Protein synonyms:
    Epidermal Growth Factor
  • Species:
    Rat

Specifications